missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Furanylmethylene)malononitrile, 98+%, Thermo Scientific™
£18.38 - £47.17
Identificadores de productos químicos
CAS | 3237-22-7 |
---|---|
Molecular Formula | C8H4N2O |
Molecular Weight (g/mol) | 144.13 |
MDL Number | MFCD00137623 |
InChI Key | CRUJHLHRJSNZIU-UHFFFAOYSA-N |
Synonym | furfurylidenemalononitrile, 2-furan-2-ylmethylene malononitrile, furfural-malonitril, 2-furanylmethylene malononitrile, malononitrile, furfurylidene, 2-furan-2-ylmethylidene propanedinitrile, 2-furfurylidenemalononitrile, furfural-malonitril german, 1,1-dicyano-2-2-furyl ethylene, furan-2-ylmethylidene propanedinitrile |
PubChem CID | 18606 |
SMILES | N#CC(=CC1=CC=CO1)C#N |
Descripción
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
3237-22-7 | |
144.13 | |
CRUJHLHRJSNZIU-UHFFFAOYSA-N | |
18606 |
C8H4N2O | |
MFCD00137623 | |
furfurylidenemalononitrile, 2-furan-2-ylmethylene malononitrile, furfural-malonitril, 2-furanylmethylene malononitrile, malononitrile, furfurylidene, 2-furan-2-ylmethylidene propanedinitrile, 2-furfurylidenemalononitrile, furfural-malonitril german, 1,1-dicyano-2-2-furyl ethylene, furan-2-ylmethylidene propanedinitrile | |
N#CC(=CC1=CC=CO1)C#N |
Especificaciones
3237-22-7 | |
98% min. (GC) | |
MFCD00137623 | |
CRUJHLHRJSNZIU-UHFFFAOYSA-N | |
2-(furan-2-ylmethylidene)propanedinitrile | |
18606 | |
98+% |
70°C to 74°C | |
C8H4N2O | |
furfurylidenemalononitrile, 2-furan-2-ylmethylene malononitrile, furfural-malonitril, 2-furanylmethylene malononitrile, malononitrile, furfurylidene, 2-furan-2-ylmethylidene propanedinitrile, 2-furfurylidenemalononitrile, furfural-malonitril german, 1,1-dicyano-2-2-furyl ethylene, furan-2-ylmethylidene propanedinitrile | |
N#CC(=CC1=CC=CO1)C#N | |
144.13 | |
114.13 | |
(2-Furanylmethylene)malononitrile |
Seguridad y manipulación
GHS Signal Word: Warning
Nombre del producto
Palabra de advertencia
Clasificación
Indicaciones de peligro
Consejos de prudencia
- (2-Furanylmethylene)malononitrile
Palabra de advertencia
- Warning
Clasificación
- Acute toxicity Category 4
Indicaciones de peligro
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Consejos de prudencia
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only