missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

£43.90 - £476.00

Chemical Identifiers

CAS 672-13-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00003332
InChI Key FZHSPPYCNDYIKD-UHFFFAOYSA-N
Synonym 5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649
PubChem CID 95695
IUPAC Name 2-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)O)C=O
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11482398
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15753.06
5 g
£43.90
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11492398
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15753.14
25 g
£145.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11319609
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15753.18
50 g
£274.00
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11402408
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15753.22
100 g
£476.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.

Solubility
Miscible with chloroform. Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

672-13-9
152.149
FZHSPPYCNDYIKD-UHFFFAOYSA-N
95695
COC1=CC(=C(C=C1)O)C=O
C8H8O3
MFCD00003332
5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649
2-hydroxy-5-methoxybenzaldehyde

Specifications

672-13-9
1.221
>110°C (230°F)
1.578
1100532
5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649
FZHSPPYCNDYIKD-UHFFFAOYSA-N
2-hydroxy-5-methoxybenzaldehyde
95695
98%
13°C to 16°C
248°C to 252°C
C8H8O3
MFCD00003332
Air Sensitive
Miscible with chloroform. Slightly miscible with water.
COC1=CC(=C(C=C1)O)C=O
152.149
152.15
2-Hydroxy-5-methoxybenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 211-589-0

missing translation for 'rtecsNumber' : BZ2800000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only