Learn More
2-Mercaptobenzyl alcohol, tech. 90%, Thermo Scientific Chemicals
£29.60 - £365.00
Chemical Identifiers
CAS | 4521-31-7 |
---|---|
Molecular Formula | C7H8OS |
Molecular Weight (g/mol) | 140.2 |
MDL Number | MFCD00014448 |
InChI Key | FYWFCRHZXORPFH-UHFFFAOYSA-N |
Synonym | 2-mercaptobenzyl alcohol, 2-mercaptophenyl methanol, o-mercaptobenzyl alcohol, 2-sulfanylphenyl methanol, 2-mercaptobenzylalcohol, 2-hydroxymethyl thiophenol, benzenemethanol, 2-mercapto, hydroxymethylthiophenol, o-mercaptobenzylalcohol, acmc-1csoz |
PubChem CID | 138278 |
IUPAC Name | (2-sulfanylphenyl)methanol |
SMILES | C1=CC=C(C(=C1)CO)S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322477
|
Thermo Scientific Alfa Aesar
L11536.03 |
1 g |
£29.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332477
|
Thermo Scientific Alfa Aesar
L11536.06 |
5 g |
£103.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11342477
|
Thermo Scientific Alfa Aesar
L11536.14 |
25 g |
£365.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4521-31-7 | |
140.2 | |
FYWFCRHZXORPFH-UHFFFAOYSA-N | |
138278 | |
C1=CC=C(C(=C1)CO)S |
C7H8OS | |
MFCD00014448 | |
2-mercaptobenzyl alcohol, 2-mercaptophenyl methanol, o-mercaptobenzyl alcohol, 2-sulfanylphenyl methanol, 2-mercaptobenzylalcohol, 2-hydroxymethyl thiophenol, benzenemethanol, 2-mercapto, hydroxymethylthiophenol, o-mercaptobenzylalcohol, acmc-1csoz | |
(2-sulfanylphenyl)methanol |
Specifications
4521-31-7 | |
1.21 | |
>110°C (230°F) | |
C7H8OS | |
MFCD00014448 | |
2-mercaptobenzyl alcohol, 2-mercaptophenyl methanol, o-mercaptobenzyl alcohol, 2-sulfanylphenyl methanol, 2-mercaptobenzylalcohol, 2-hydroxymethyl thiophenol, benzenemethanol, 2-mercapto, hydroxymethylthiophenol, o-mercaptobenzylalcohol, acmc-1csoz | |
C1=CC=C(C(=C1)CO)S | |
140.2 | |
140.21 | |
Technical |
31°C to 32°C | |
94°C to 96°C (0.2 mmHg) | |
Like Rotten Eggs | |
1.623 | |
2206253 | |
FYWFCRHZXORPFH-UHFFFAOYSA-N | |
(2-sulfanylphenyl)methanol | |
138278 | |
90% | |
2-Mercaptobenzyl alcohol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P280-P305+P351+P338-P310
H318
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only