Learn More
2-Methylbutane, 99+%, for spectroscopy, Thermo Scientific Chemicals
£9.30 - £55.20
Chemical Identifiers
CAS | 78-78-4 |
---|---|
Molecular Formula | C5H12 |
Molecular Weight (g/mol) | 72.15 |
MDL Number | MFCD00009338 |
InChI Key | QWTDNUCVQCZILF-UHFFFAOYSA-N |
Synonym | isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 |
PubChem CID | 6556 |
ChEBI | CHEBI:30362 |
IUPAC Name | 2-methylbutane |
SMILES | CCC(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10665222
|
Thermo Scientific Acros
167840250 |
25 mL | Glass bottle | ||||||
10255740
|
Thermo Scientific Acros
167842500 |
250 mL | Glass bottle |
£24.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10407010
|
Thermo Scientific Acros
167840010 |
1 L | Glass bottle |
£55.20
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopyChemical Identifiers
78-78-4 | |
72.15 | |
QWTDNUCVQCZILF-UHFFFAOYSA-N | |
6556 | |
2-methylbutane |
C5H12 | |
MFCD00009338 | |
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
CHEBI:30362 | |
CCC(C)C |
Specifications
0.0005 meq/g max. | |
(1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 max. at 240nm, 0.05 max. at 230nm, 0.07 max. at 220nm, 0.3 max. at 210nm, 1 max. at 192nm | |
100.0 | |
0.6200g/mL | |
−51°C | |
C5H12 | |
1.3530 to 1.3550 | |
01, 134 | |
0.62 | |
Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons, oils and ether, slightly soluble in alcohol | |
CCC(C)C | |
72.15 | |
CHEBI:30362 | |
99+% | |
Liquid |
78-78-4 | |
99.0 | |
-160.0°C | |
30.0°C | |
99% min. (GC) | |
C2H5CH(CH3)2 | |
MFCD00009338 | |
0.0003% max. | |
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
QWTDNUCVQCZILF-UHFFFAOYSA-N | |
2-methylbutane | |
6556 | |
72.15 | |
Spectroscopy | |
2-Methylbutane, 99+% |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
Repeated exposure may cause skin drynes
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Keep away from heat/sparks/open flam
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-142-8
missing translation for 'rtecsNumber' : EK4430000
missing translation for 'tsca' : TSCA
- 2-Methylbutane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H224-Extremely flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
RUO – Research Use Only