Learn More
2-Methylcyclohexanone, 98%, Thermo Scientific Chemicals
£29.75 - £365.82
Chemical Identifiers
CAS | 583-60-8 |
---|---|
InChI Key | LFSAPCRASZRSKS-UHFFFAOYSA-N |
Synonym | 2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone |
PubChem CID | 11419 |
IUPAC Name | 2-methylcyclohexan-1-one |
SMILES | CC1CCCCC1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10551462
|
Thermo Scientific Acros
126610500 |
50 g |
£29.75
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10561462
|
Thermo Scientific Acros
126612500 |
250 g |
£97.59
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10638124
|
Thermo Scientific Acros
126610010 |
1 kg |
£365.82
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
583-60-8 | |
2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone | |
2-methylcyclohexan-1-one |
LFSAPCRASZRSKS-UHFFFAOYSA-N | |
11419 | |
CC1CCCCC1=O |
Specifications
583-60-8 | |
100.0 | |
Colorless to Yellow | |
162.0°C to 163.0°C | |
Authentic | |
Glass bottle | |
CH3C6H9(=O) | |
07, 14 | |
2-methylcyclohexanone, o-methylcyclohexanone, cyclohexanone, 2-methyl, 2-metilcicloesanone, 2-methyl-1-cyclohexanone, methylanon, alpha-methylcyclohexanone, sexton b, 2-methyl-cyclohexanon, 2-methyl cyclohexanone | |
LFSAPCRASZRSKS-UHFFFAOYSA-N | |
2-methylcyclohexan-1-one | |
11419 | |
98% | |
2-Methylcyclohexanone, 98% |
97.5 | |
-14.0°C | |
0.9240g/mL | |
48°C | |
97.5% min. (GC) | |
C7H12O | |
1.4470 to 1.4490 | |
0.924 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
CC1CCCCC1=O | |
112.17 | |
112.17 | |
Liquid |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-513-6
missing translation for 'rtecsNumber' : GW1750000
missing translation for 'tsca' : TSCA
RUO – Research Use Only