missing translation for 'onlineSavingsMsg'
Learn More
2-Nitro-p-phenylenediamine, 95%, Thermo Scientific Chemicals Product Code.: 11445477

2-Nitro-p-phenylenediamine, 95%, Thermo Scientific Chemicals

Product Code. 11445477
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11445477

Brand: Thermo Scientific Alfa Aesar B23477.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5307-14-2
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00007903
InChI Key HVHNMNGARPCGGD-UHFFFAOYSA-N
Synonym 2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2rShow MoreShow Less
PubChem CID 4338370
ChEBI CHEBI:76394
IUPAC Name 2-nitrobenzene-1,4-diamine
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])N

Specifications

Melting Point 134°C to 137°C
Quantity 100 g
Formula Weight 153.14
Percent Purity 95%
Chemical Name or Material 2-Nitro-p-phenylenediamine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243260 Certificate TypeCertificate of Analysis Date07/09/2022 Catalog NumberB23477.22,B23477.14,B23477.36
Lot Number10236651 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23477.22,B23477.14,B23477.36
Lot Number10133570 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23477.22,B23477.14,B23477.36
Lot NumberH1136A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23477.22,B23477.14,B23477.36
Lot Number10149773 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23477.22,B23477.14,B23477.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Nitro-p-phenylenediamine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.