missing translation for 'onlineSavingsMsg'
Learn More
2-Pentyn-1-ol, 98%, Thermo Scientific Chemicals Product Code.: 11439893

2-Pentyn-1-ol, 98%, Thermo Scientific Chemicals

Product Code. 11439893
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11439893

Brand: Thermo Scientific Alfa Aesar A12999.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6261-22-9
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00040915
InChI Key WLPYSOCRPHTIDZ-UHFFFAOYSA-N
Synonym 2-pentyn-1-ol, 2-pentyne-1-ol, 2-pentynol, zlchem 1280, pubchem13063, acmc-209n5w, ksc357i0r, 2-pentyn-1-01
PubChem CID 80421
IUPAC Name pent-2-yn-1-ol
SMILES CCC#CCO

Specifications

Density 0.91
Boiling Point 154°C
Flash Point 58°C (136°F)
Odor Recognizable
Linear Formula CH3CH2C≡CCH2OH
Refractive Index 1.453
Quantity 1 g
UN Number UN1987
Beilstein 1734007
Formula Weight 84.12
Percent Purity 98%
Chemical Name or Material 2-Pentyn-1-ol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Pentyn-1-ol, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.