missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1-propylboronic acid pinacol ester, 97%, Thermo Scientific™

£65.38 - £609.00

Chemical Identifiers

CAS 280559-30-0
Molecular Formula C15H23BO2
Molecular Weight (g/mol) 246.157
MDL Number MFCD09953475
InChI Key GTZNZJQZUWIDTN-UHFFFAOYSA-N
Synonym 2-phenyl-1-propylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-phenylpropyl-1,3,2-dioxaborolane, 2-phenylpropylboronic acid pinacol ester, 2-2-phenylpropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 46739058
IUPAC Name 4,4,5,5-tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CC(C)C2=CC=CC=C2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15467635
View Documents
Thermo Scientific Alfa Aesar
H26931.MD
250 mg
£65.38
250mg
Please to purchase this item. Need a web account? Register with us today!
11491510
View Documents
Thermo Scientific Alfa Aesar
H26931.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11401520
View Documents
Thermo Scientific Alfa Aesar
H26931.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

280559-30-0
246.157
GTZNZJQZUWIDTN-UHFFFAOYSA-N
46739058
B1(OC(C(O1)(C)C)(C)C)CC(C)C2=CC=CC=C2
C15H23BO2
MFCD09953475
2-phenyl-1-propylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-phenylpropyl-1,3,2-dioxaborolane, 2-phenylpropylboronic acid pinacol ester, 2-2-phenylpropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
4,4,5,5-tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane

Specifications

280559-30-0
MFCD09953475
GTZNZJQZUWIDTN-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane
46739058
97%
C15H23BO2
2-phenyl-1-propylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-phenylpropyl-1,3,2-dioxaborolane, 2-phenylpropylboronic acid pinacol ester, 2-2-phenylpropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)CC(C)C2=CC=CC=C2
246.157
246.16
2-Phenyl-1-propylboronic acid pinacol ester
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.