missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylcyclohexanone, 98%, Thermo Scientific Chemicals
£32.36 - £300.50
Chemical Identifiers
CAS | 1444-65-1 |
---|---|
Molecular Formula | C12H14O |
Molecular Weight (g/mol) | 174.243 |
MDL Number | MFCD00001629 |
InChI Key | DRLVMOAWNVOSPE-UHFFFAOYSA-N |
Synonym | 2-phenylcyclohexanone, cyclohexanone, 2-phenyl, 2-phenyl-cyclohexanone, 2-phenyl cyclohexanone, cyclohexanone, phenyl, 2-phenylcyclohexanon, phenylcyclohexanone, phenyl cyclohexanone, 6-phenylcyclohexanone |
PubChem CID | 95592 |
IUPAC Name | 2-phenylcyclohexan-1-one |
SMILES | C1CCC(=O)C(C1)C2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11473577
|
Thermo Scientific Alfa Aesar
A17340.03 |
1 g |
£32.36
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11483577
|
Thermo Scientific Alfa Aesar
A17340.06 |
5 g |
£93.47
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493577
|
Thermo Scientific Alfa Aesar
A17340.14 |
25 g |
£300.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It can be used to produce 6-oxo-6-phenyl-hexanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt can be used to produce 6-oxo-6-phenyl-hexanoic acid.
Solubility
Slightly soluble in water.
Notes
Storage temperature: 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
1444-65-1 | |
174.243 | |
DRLVMOAWNVOSPE-UHFFFAOYSA-N | |
95592 | |
C1CCC(=O)C(C1)C2=CC=CC=C2 |
C12H14O | |
MFCD00001629 | |
2-phenylcyclohexanone, cyclohexanone, 2-phenyl, 2-phenyl-cyclohexanone, 2-phenyl cyclohexanone, cyclohexanone, phenyl, 2-phenylcyclohexanon, phenylcyclohexanone, phenyl cyclohexanone, 6-phenylcyclohexanone | |
2-phenylcyclohexan-1-one |
Specifications
1444-65-1 | |
C12H14O | |
2-phenylcyclohexanone, cyclohexanone, 2-phenyl, 2-phenyl-cyclohexanone, 2-phenyl cyclohexanone, cyclohexanone, phenyl, 2-phenylcyclohexanon, phenylcyclohexanone, phenyl cyclohexanone, 6-phenylcyclohexanone | |
DRLVMOAWNVOSPE-UHFFFAOYSA-N | |
2-phenylcyclohexan-1-one | |
95592 | |
98% |
56°C to 59°C | |
MFCD00001629 | |
Slightly soluble in water. | |
C1CCC(=O)C(C1)C2=CC=CC=C2 | |
174.243 | |
174.24 | |
2-Phenylcyclohexanone |
Safety and Handling
missing translation for 'einecsNumber' : 215-888-7
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only