missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol CHROMASOLV™, for HPLC, 99.9%, Honeywell Riedel-de Haën™
CHROMASOLV™, for HPLC, 99.9%
£23.59 - £3618.24
Chemical Identifiers
CAS | 67-63-0 |
---|---|
Molecular Formula | C3H8O |
Molecular Weight (g/mol) | 60.096 |
MDL Number | MFCD00011674 |
InChI Key | KFZMGEQAYNKOFK-UHFFFAOYSA-N |
Synonym | isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
PubChem CID | 3776 |
ChEBI | CHEBI:17824 |
IUPAC Name | propan-2-ol |
SMILES | CC(C)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15664590
|
Honeywell-Chromasolv
34863-100ML |
100 mL | N/A | ||||||
15684590
|
Honeywell-Chromasolv
34863-1L |
1 L |
£48.38
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15614600
|
Honeywell-Chromasolv
34863-2L |
2 L | N/A | ||||||
15694590
|
Honeywell-Chromasolv
34863-2.5L |
2.5 L |
£96.43
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15604600
|
Honeywell-Chromasolv
34863-2.5L-PC |
2.5 L-PC | N/A | ||||||
15634600
|
Honeywell-Chromasolv
34863-6X1L |
6 x 1 L |
£209.76
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15624600
|
Honeywell-Chromasolv
34863-4X2.5L |
4 x 2.5 L |
£322.76
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15644600
|
Honeywell Riedel-de Haën
34863-72X2.5L |
72 x 2.5 L | N/A | ||||||
15674590
|
Honeywell-Chromasolv
34863-185L |
185 L |
£3618.24
185 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
67-63-0 | |
60.096 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
3776 | |
propan-2-ol |
C3H8O | |
MFCD00011674 | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CHEBI:17824 | |
CC(C)O |
Specifications
-89.5°C | |
0.785g/mL at 25°C | |
n20/D 1.377 | |
(CH3)2CHOH | |
44mmHg (25°C) | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol | |
CC(C)O | |
60.096 | |
CHEBI:17824 | |
99.9% | |
2-Propanol, for HPLC |
67-63-0 | |
82°C | |
C3H8O | |
MFCD00011674 | |
635639 | |
KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
propan-2-ol | |
3776 | |
60.1g/mol | |
CHROMASOLV™ |
Safety and Handling
P210-P261-P305 + P351 + P338
H225-H319-H336
missing translation for 'einecsNumber' : 200-661-7
missing translation for 'rtecsNumber' : NT8050000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Propanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title