missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, puriss., Honeywell
BIOPHARMA

£31.03 - £2046.24

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 8
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15612520
View Documents
Honeywell Riedel-de Haën
24137-100ML
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15652520
BIOPHARMA
View Documents
Honeywell Riedel-de Haën
24137-1L
1 L
£31.03
1 L
Please to purchase this item. Need a web account? Register with us today!
15662520
BIOPHARMA
View Documents Promotion Details
Honeywell Riedel-de Haën
24137-2.5L
2.5 L
£70.10
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15612530
BIOPHARMA
View Documents Promotion Details
Honeywell Riedel-de Haën
24137-5L
5 L
£108.34
5 L
Please to purchase this item. Need a web account? Register with us today!
15622520
View Documents
Honeywell Riedel-de Haën
24137-10L
10 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15682520
BIOPHARMA
View Documents
Honeywell Riedel-de Haën
24137-25L
25 L
£514.10
25 L
Please to purchase this item. Need a web account? Register with us today!
15672520
BIOPHARMA
View Documents
Honeywell Riedel-de Haën
24137-20KG
20 kg
£1046.64
20kg
Please to purchase this item. Need a web account? Register with us today!
15642520
BIOPHARMA
View Documents
Honeywell Riedel-de Haën
24137-160KG
160 kg
£2046.24
160kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

67-63-0
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
propan-2-ol
C3H8O
MFCD00011674
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CHEBI:17824
CC(C)O

Specifications

67-63-0
0.02
750°F
(CH3)2CHOH
33mmHg (20°C), 44mmHg (25°C)
635639
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1g/mol
puriss.
0.785g/mL at 25°C
12°C (53.6°F)
C3H8O
MFCD00011674
1219
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.096
CHEBI:17824
≥99.5% (GC)
2-Propanol, Meets analytical specification of Ph. Eur.BP, USP
Videos
Safety and Handling

Safety and Handling

P210-P261-P305 + P351 + P338

H225-H319-H336

Danger

missing translation for 'einecsNumber' : 200-661-7

missing translation for 'rtecsNumber' : NT8050000

Product Identifier
  • 2-Propanol

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.