missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridineboronic acid N-phenyl-diethanolamine ester, 50-70%, see CofA for exact com, Thermo Scientific Chemicals

£28.60 - £155.00

Chemical Identifiers

CAS 662138-96-7, 67-63-0, 120-07-0
Molecular Formula C15H17BN2O2
Molecular Weight (g/mol) 268.12
MDL Number MFCD06798116
InChI Key FZYLTWAWSRCZPG-UHFFFAOYSA-N
Synonym 2-pyridineboronic acid 2,2'-phenylimino diethanol ester, pyridine-2-boronic acid n-phenyldiethanolamine ester x isopropanol, pubchem6434, 6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane; isopropyl alcohol
PubChem CID 49761666
SMILES C1C[N+]2(CCO[B-]2(O1)C1=NC=CC=C1)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11966001
View Documents
Thermo Scientific Acros
439890010
1 g Glass bottle
£28.60
1g
Please to purchase this item. Need a web account? Register with us today!
11986001
View Documents
Thermo Scientific Acros
439890050
5 g Glass bottle
£83.50
5g
Please to purchase this item. Need a web account? Register with us today!
11996001
View Documents
Thermo Scientific Acros
439890100
10 g Glass bottle
£155.00
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

662138-96-7, 67-63-0, 120-07-0
268.12
FZYLTWAWSRCZPG-UHFFFAOYSA-N
49761666
C15H17BN2O2
MFCD06798116
2-pyridineboronic acid 2,2'-phenylimino diethanol ester, pyridine-2-boronic acid n-phenyldiethanolamine ester x isopropanol, pubchem6434, 6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane; isopropyl alcohol
C1C[N+]2(CCO[B-]2(O1)C1=NC=CC=C1)C1=CC=CC=C1

Specifications

662138-96-7, 67-63-0, 120-07-0
White
C15H17BN2O2
2-pyridineboronic acid 2,2'-phenylimino diethanol ester, pyridine-2-boronic acid n-phenyldiethanolamine ester x isopropanol, pubchem6434, 6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane; isopropyl alcohol
C1C[N+]2(CCO[B-]2(O1)C1=NC=CC=C1)C1=CC=CC=C1
268.12
268.13
Contains varying amounts of IPA and N-phenyldiethanolamine
2-Pyridineboronic acid N-phenyl-diethanolamine ester
>300°C
Authentic
MFCD06798116
FZYLTWAWSRCZPG-UHFFFAOYSA-N
6-phenyl-2-pyridin-2-yl-1,3,6,2-dioxazaborocane;propan-2-ol
49761666
50-70%,see CofA for exact com
Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves

GHS Signal Word: Warning

Product Identifier
  • 2-Pyridineboronic acid N-phenyldiethanolamine ester

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.