Learn More
2-Pyrrolidin-1-ylisonicotinic acid, ≥90%, Thermo Scientific™
£38.00 - £70.20
Chemical Identifiers
CAS | 98088-04-1 |
---|---|
Molecular Formula | C10H12N2O2 |
Molecular Weight (g/mol) | 192.22 |
MDL Number | MFCD04115212 |
InChI Key | PQLHQGBNMJTAEH-UHFFFAOYSA-N |
Synonym | 2-pyrrolidin-1-yl-isonicotinic acid, 2-pyrrolidin-1-yl isonicotinic acid, 2-pyrrolidin-1-yl pyridine-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-1-pyrrolidinyl, 2-pyrrolidin-1-ylisonicotinic acid, 2-1-pyrrolidinyl isonicotinic acid, 2-1-pyrrolidinyl-4-pyridinecarboxylic acid, 2-pyrrolidin-1-ylisonicotinic acid, 1.5 hydrate, 2-pyrrolidinylpyridine-4-carboxylic acid, hydrochloride |
PubChem CID | 6484236 |
IUPAC Name | 2-pyrrolidin-1-ylpyridine-4-carboxylic acid |
SMILES | OC(=O)C1=CC(=NC=C1)N1CCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11350677
|
Thermo Scientific Maybridge
CC50401CB |
250 mg | Amber Glass Bottle |
£38.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10670454
|
Thermo Scientific Maybridge
CC50401DA |
1 g | Amber Glass Bottle |
£70.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
98088-04-1 | |
192.22 | |
PQLHQGBNMJTAEH-UHFFFAOYSA-N | |
6484236 | |
OC(=O)C1=CC(=NC=C1)N1CCCC1 |
C10H12N2O2 | |
MFCD04115212 | |
2-pyrrolidin-1-yl-isonicotinic acid, 2-pyrrolidin-1-yl isonicotinic acid, 2-pyrrolidin-1-yl pyridine-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-1-pyrrolidinyl, 2-pyrrolidin-1-ylisonicotinic acid, 2-1-pyrrolidinyl isonicotinic acid, 2-1-pyrrolidinyl-4-pyridinecarboxylic acid, 2-pyrrolidin-1-ylisonicotinic acid, 1.5 hydrate, 2-pyrrolidinylpyridine-4-carboxylic acid, hydrochloride | |
2-pyrrolidin-1-ylpyridine-4-carboxylic acid |
Specifications
98088-04-1 | |
100.0 | |
C10H12N2O2 | |
2-pyrrolidin-1-yl-isonicotinic acid, 2-pyrrolidin-1-yl isonicotinic acid, 2-pyrrolidin-1-yl pyridine-4-carboxylic acid, 4-pyridinecarboxylic acid, 2-1-pyrrolidinyl, 2-pyrrolidin-1-ylisonicotinic acid, 2-1-pyrrolidinyl isonicotinic acid, 2-1-pyrrolidinyl-4-pyridinecarboxylic acid, 2-pyrrolidin-1-ylisonicotinic acid, 1.5 hydrate, 2-pyrrolidinylpyridine-4-carboxylic acid, hydrochloride | |
OC(=O)C1=CC(=NC=C1)N1CCCC1 | |
192.22 | |
219.25 | |
2-Pyrrolidin-1-ylisonicotinic acid, 1.5 hydrate |
89.5 | |
90% | |
MFCD04115212 | |
PQLHQGBNMJTAEH-UHFFFAOYSA-N | |
2-pyrrolidin-1-ylpyridine-4-carboxylic acid | |
6484236 | |
90% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning