Learn More
2-(Trifluoromethyl)pyrrolidine, 97%, Thermo Scientific Chemicals
£132.35 - £478.00
Chemical Identifiers
CAS | 109074-67-1 |
---|---|
Molecular Formula | C5H8F3N |
Molecular Weight (g/mol) | 139.12 |
InChI Key | VINAMCOZNJHNIH-UHFFFAOYSA-N |
Synonym | 2-trifluoromethyl pyrrolidine, 2-trifluoromethyl-pyrrolidine, dl-2-trifluoromethylpyrrolidine, pyrrolidine,2-trifluoromethyl, pyrrolidine, 2-trifluoromethyl, +/--2-trifluoromethyl pyrrolidine, acmc-20dptq, acmc-1cih7, 2-trifluoromethyl tetrahydropyrrole |
PubChem CID | 2782839 |
IUPAC Name | 2-(trifluoromethyl)pyrrolidine |
SMILES | C1CC(NC1)C(F)(F)F |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109074-67-1 | |
139.12 | |
2-trifluoromethyl pyrrolidine, 2-trifluoromethyl-pyrrolidine, dl-2-trifluoromethylpyrrolidine, pyrrolidine,2-trifluoromethyl, pyrrolidine, 2-trifluoromethyl, +/--2-trifluoromethyl pyrrolidine, acmc-20dptq, acmc-1cih7, 2-trifluoromethyl tetrahydropyrrole | |
2-(trifluoromethyl)pyrrolidine |
C5H8F3N | |
VINAMCOZNJHNIH-UHFFFAOYSA-N | |
2782839 | |
C1CC(NC1)C(F)(F)F |
Specifications
109074-67-1 | |
110.0°C to 112.0°C | |
Authentic | |
Glass Bottle | |
1.21 | |
VINAMCOZNJHNIH-UHFFFAOYSA-N | |
2-(trifluoromethyl)pyrrolidine | |
2782839 | |
97% |
1.2100g/mL | |
26°C | |
96% min. (GC) | |
C5H8F3N | |
2-trifluoromethyl pyrrolidine, 2-trifluoromethyl-pyrrolidine, dl-2-trifluoromethylpyrrolidine, pyrrolidine,2-trifluoromethyl, pyrrolidine, 2-trifluoromethyl, +/--2-trifluoromethyl pyrrolidine, acmc-20dptq, acmc-1cih7, 2-trifluoromethyl tetrahydropyrrole | |
C1CC(NC1)C(F)(F)F | |
139.12 | |
139.12 | |
2-(Trifluoromethyl)pyrrolidine |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to
GHS Signal Word: Warning
- 2-(Trifluoromethyl)pyrrolidine, 97%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only