Learn More
uvex 2124 Xact-Fit Multi Reusable Ear Plugs with Storage Case
Innovative, corded ear plugs feature a unique oval design that adjusts perfectly to the anatomy of the ear canal. The angled pin significantly reduces pressure in the ear, while remaining comfortable to wear for long periods of time.
Brand:  uvex 2124018
71.17 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 17200352
  • £90.80 / Pack of 50
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unique oval design adjusts perfectly to the anatomy of the ear canal
  • Angled pin significantly reduces pressure in the ear
  • Pre-formed pins make it easy to insert and remove ear plugs
  • Thumb indentations for enable quick and easy handling
  • Smooth, dirt-repellent surface can be cleaned with mild soap and water
  • Comfortably wear corded ear plugs around the neck in low-noise areas
  • Clear, hygienic box included for safe storage when not in use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transparent, Gray
Pocket-size case
Regular
2124 Xact-Fit Multi
Corded
Ear Plugs
No
EN 352-2
No
Lime Green
Variable Environments
26 dB
Oval
Reusable
No