missing translation for 'onlineSavingsMsg'
Learn More

Oxoid 24 LEB Selective Supplement

The 24 LEB selective supplement from Oxoid includes ten vials. One vial is required for 500 mL of medium.

Brand:  Oxoid SR0243E

Product Code. 13245419

  • £115.00 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The 24 LEB selective supplement from Oxoid includes ten vials. One vial is required for 500 mL of medium.

  • Selectively enrich Listeria monocytogenes and other Listeria spp. from food and environmental samples
  • Medium provides optimal growing conditions for all Listeria species for PCR applications
  • Specifically designed for use with PCR systems, such as Thermo Scientific™ SureTect™ molecular assays for Listeria

Directions:

  • Completely dissolve 21.75 g of 24 LEB base per 500 mL of distilled water
  • Sterilize by autoclaving at 121 °C for 15 min.
  • Cool to 50 °C
  • Aseptically add contents of one vial of 24 LEB selective supplement per 500 mL medium, reconstituted as directed
  • Cool to room temp before use, once made, store medium at 2 to 8 °C in dark

Technique:

  • Refer to BAX™ system user guide and product inserts for PCR Assay L. monocytogenes 24E (QB8125C) and BAX™ system PCR Assay Genus Listeria 24E (QB8135C) for instructions for use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Listeria
Adheres to ISO 11290-1:1997 standards
United Kingdom
Selective enrichment of Listeria sp from food and environmental samples
10 Pack
Yellow/orange pellet
Room Temperature
Dehydrated culture media
Tub
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6224772 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog NumberSR0243E
Lot Number6220651 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog NumberSR0243E
Lot Number6208464 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberSR0243E
Lot Number6205298 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberSR0243E
Lot Number6171443 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog NumberSR0243E
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Oxoid 24 LEB Selective Supplement > 24 LEB Selective Supplement

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.