Learn More
25-Hydroxy VD2-d6, MedChemExpress
MedChemExpress 25-Hydroxy VD2-d6 is the deuterium labeled 25-Hydroxy VD2.
Brand: Medchem Express HY-15328
This item is not returnable.
View return policy
Description
25-Hydroxy VD2-d6 is the deuterium labeled 25-Hydroxy VD2.Specifications
25-Hydroxy VD2-d6 | |
5 mg | |
Research | |
DMSO : 100 mg/mL (238.85 mM; Need ultrasonic) | |
418.68 | |
98.96% | |
Solid |
1262843-46-8 | |
White | |
C28H38D6O2 | |
O[C@@H](CCC/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](/C=C/[C@H](C)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)([H])[C@]3(C)CCC\2 | |
418.68 | |
Research |