Learn More
25-Hydroxy VD2-d6, MedChemExpress
MedChemExpress 25-Hydroxy VD2-d6 is the deuterium labeled 25-Hydroxy VD2.
£1052.70 - £3661.46
Specifications
Chemical Name or Material | 25-Hydroxy VD2-d6 |
---|---|
CAS | 1262843-46-8 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C28H38D6O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16439047
|
Medchem Express
HY-15328 |
5 mg |
£3661.46
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16401509
|
Medchem Express
HY-15328 |
1 mg |
£1052.70
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
25-Hydroxy VD2-d6 is the deuterium labeled 25-Hydroxy VD2.Specifications
25-Hydroxy VD2-d6 | |
White | |
C28H38D6O2 | |
O[C@@H](CCC/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](/C=C/[C@H](C)C(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)([H])[C@]3(C)CCC\2 | |
418.68 | |
Research |
1262843-46-8 | |
Research | |
DMSO : 100 mg/mL (238.85 mM; Need ultrasonic) | |
418.68 | |
98.96% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : 4°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
Recommended Storage : 4°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.