missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 25-Hydroxycholesterol
Antiviral; also α-crystallin (cryAB) chaperone
Brand: Tocris Bioscience™ 5741/10
82.94 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
25-Hydroxycholesterol is an antiviral. Stimulates expression of miR-185 microRNA which inhibits the hepatitis C virus life cycle via inhibition of cholesterol biosynthesis and virus cell membrane fusion. Also α-crystallin (cryAB) chaperone (Kd = 10.1 μM). Blocks α-crystallin aggregation and partially reverses cataract formation by binding and stabilizing soluble forms of cryAB in vitro and in vivo.Specifications
Cholest-5-ene-3β,25-diol | |
2140-46-7 | |
402.65 | |
Antiviral Compounds |
Inhibitor | |
C27H46O2 | |
Store at -20°C | |
10 mg |
For Research Use Only