Learn More
Hamilton™ 25 μL 1700 Series Gas Tight Syringe
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
Brand: Hamilton™ CAL80275
This item is not returnable.
View return policy
Description
- This series of syringes are ideal for dispensing volumes up to 25 μL.
- A variety of different terminations and needle options are available in this series.
Specifications
Not Autoclavable | |
22s G | |
0.0059 in. | |
0.0283 in. | |
0.729 mm | |
0.72 mm | |
Gas Tight Syringe | |
50°C | |
1702 N Syringe | |
1 | |
51 mm | |
Cemented |
Borosilicate Glass | |
0.029 in. | |
0.15 mm | |
0.305 in. | |
7.75 mm | |
2 in. | |
122°F | |
50°F | |
10°C | |
25 μL | |
Cone |