Learn More
Hamilton™ 25 μL 1700 Series Gas Tight Syringe
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
Brand:  Hamilton™ CAL80230
 View more versions of this product
Product Code. 17824392
  • £363.00 / Each
Explore more special offers
Description
Description
  • This series of syringes are ideal for dispensing volumes up to 25 μL.
  • A variety of different terminations and needle options are available in this series.
Specifications
Specifications
Not Autoclavable
22s G
0.0059 in.
0.0283 in.
0.729 mm
0.72 mm
Gas Tight Syringe
50°C
1702 N
1
51 mm
Cemented
PTFE
0.029 in.
0.15 mm
0.305 in.
7.75 mm
2 in.
122°F
50°F
10°C
25 μL
Sharp, Bevel, Curved, Non-coring
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Ofertas especiales
Ofertas especiales