Learn More
Hamilton™ 25 μL 700 Series Microliter Syringe
The 700 Series Microliter Syringe fulfills the general liquid handling requirements of manufacturing and research laboratories.
Brand:  Hamilton™ CAL84855
 View more versions of this product
Product Code. 17804202
  • £400.00 / Each
Explore more special offers
Description
Description
  • The 700 series is the original Hamilton syringe.
  • This series of Microliter syringes is designed to solve the general liquid handling requirements of manufacturing and research laboratories.
  • The 700 series remains the industry standard.
  • The plungers and barrels are not interchangeable or replaceable.
  • Dispenses volumes up to 25 μL.