Learn More
Hamilton™ 25 μL 700 Series Microliter Syringe
The 700 Series Microliter Syringe fulfills the general liquid handling requirements of manufacturing and research laboratories.
£223.00 - £308.00
Product Code | Brand | Needle Gauge | Connection Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Connection Type | Price | Quantity & Availability | ||||
17854312
|
Hamilton™
CAL80465 |
No Needle Included | Luer Tip |
£223.00
Each |
Estimated Shipment: 03-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17864312
|
Hamilton™
CAL80430 |
22s G | Cemented |
£243.00
Each |
Estimated Shipment: 03-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17804202
|
Hamilton™
CAL84855 |
£308.00
Each |
Estimated Shipment: 03-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17884192
|
Hamilton™
CAL84854 |
22s G | Removable |
£297.00
Each |
Estimated Shipment: 03-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17884302
|
Hamilton™
CAL7654-01 |
No Needle Included | Removable |
£236.00
Each |
Estimated Shipment: 03-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- The 700 series is the original Hamilton syringe.
- This series of Microliter syringes is designed to solve the general liquid handling requirements of manufacturing and research laboratories.
- The 700 series remains the industry standard.
- The plungers and barrels are not interchangeable or replaceable.
- Dispenses volumes up to 25 μL.