Learn More
Hamilton™ 25 μL 700 Series Microliter Syringe
The 700 Series Microliter Syringe fulfills the general liquid handling requirements of manufacturing and research laboratories.
Brand:  Hamilton™ CAL84854
 View more versions of this product
Product Code. 17884192
  • £300.00 / Each
Explore more special offers
Description
Description
  • The 700 series is the original Hamilton syringe.
  • This series of Microliter syringes is designed to solve the general liquid handling requirements of manufacturing and research laboratories.
  • The 700 series remains the industry standard.
  • The plungers and barrels are not interchangeable or replaceable.
  • Dispenses volumes up to 25 μL.
Specifications
Specifications
Autoclavable
22s G
0.0059 in.
0.0283 in.
0.729 mm
0.72 mm
122°F
50°F
10°C
25 μL
Sharp, Bevel, Curved, Non-coring
Borosilicate Glass
0.029 in.
0.15 mm
0.305 in.
7.75 mm
2 in.
50°C
702 RN Syringe
1
51 mm
Removable