Learn More
Gibco™ 293fectin™ Transfection Reagent
293fectin™ Transfection Reagent is a proprietary, cationic lipid-based formulation for transfecting DNA into eukaryotic cells.
Brand: Gibco™ 12347500
5584.80 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
293fectin™ Transfection Reagent is a proprietary, cationic lipid-based formulation for transfecting DNA into eukaryotic cells. This reagent is optimized for transfecting suspension 293 human embryonic kidney cells (e.g., FreeStyle™ 293-F cells, Cat. No. R790-07) in defined, serum-free FreeStyle™ 293 Expression Medium (12338-018), and is intended for use with the FreeStyle™ 293 Expression System (K9000-01).
293fectin™ Transfection Reagent provides the following advantages:
- Demonstrates high transfection efficiency in suspension 293 cells and is also suitable for transfecting adherent 293 cells
- Suspension FreeStyle™ 293-F cells may be transfected in FreeStyle™ 293 Expression Medium; no medium change is required
- Add 293fectin™ reagent complexes directly to cells in culture medium
- No need to remove complexes or change or add medium following transfection
Specifications
Each unit contains one vial of reagent. Store at 4°C. 293fectin™ is guaranteed stable for 6 months when stored properly. |
|
Tube | |
Lipid-based Transfection | |
Not High Throughput-Compatible (Manual) | |
Transfection Reagent | |
Yes |
Established Cell Lines | |
Plasmid DNA | |
Transfection | |
293fectin™ | |
15 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.