missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Chloro-5-methylphenoxy)piperidine, 98%, Thermo Scientific™

£46.80 - £168.00

Chemical Identifiers

CAS 902837-30-3
Molecular Formula C12H16ClNO
Molecular Weight (g/mol) 225.716
MDL Number MFCD08061103
InChI Key GRLVAJJXUNOVRA-UHFFFAOYSA-N
Synonym 3-2-chloro-5-methylphenoxy piperidine, 3-2-chloro-5-methyl-phenoxy-piperidine, 1-chloro-4-methyl-2-3-piperidyloxy benzene
PubChem CID 24212031
IUPAC Name 3-(2-chloro-5-methylphenoxy)piperidine
SMILES CC1=CC(=C(C=C1)Cl)OC2CCCNC2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11419770
SDS View Documents
Thermo Scientific Alfa Aesar
H50991.MD
250 mg N/A
N/A
11429770
SDS View Documents
Thermo Scientific Alfa Aesar
H50991.03
1 g N/A
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(2-Chloro-5-methylphenoxy)piperidine, 98%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.