missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Methyl-3-furylthio)-4-heptanone, 97%, Thermo Scientific™
£16.68 - £44.60
Chemical Identifiers
CAS | 61295-41-8 |
---|---|
Molecular Formula | C12H18O2S |
Molecular Weight (g/mol) | 226.33 |
MDL Number | MFCD01723735 |
InChI Key | GFRRQSASJZMMJC-UHFFFAOYNA-N |
Synonym | 3-2-methyl-3-furylthio-4-heptanone, 3-2-methylfuran-3-yl thio heptan-4-one, fema no. 3570, 3-2-methyl-3-furyl thio-4-heptanone, 1,3-diethylacetonyl 3-methyl-3-furyl sulfide, 3-2-methyl-3-furyl thio heptan-4-one, 4-heptanone, 3-2-methyl-3-furanyl thio, 4-heptanone, 3-2-methyl-3-furyl thio, 3-2-methylfuran-3-yl sulfanylheptan-4-one, 3-2-methyl-3-furanyl thio-4-heptanone |
PubChem CID | 43598 |
IUPAC Name | 3-(2-methylfuran-3-yl)sulfanylheptan-4-one |
SMILES | CCCC(=O)C(CC)SC1=C(C)OC=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
61295-41-8 | |
226.33 | |
GFRRQSASJZMMJC-UHFFFAOYNA-N | |
43598 | |
CCCC(=O)C(CC)SC1=C(C)OC=C1 |
C12H18O2S | |
MFCD01723735 | |
3-2-methyl-3-furylthio-4-heptanone, 3-2-methylfuran-3-yl thio heptan-4-one, fema no. 3570, 3-2-methyl-3-furyl thio-4-heptanone, 1,3-diethylacetonyl 3-methyl-3-furyl sulfide, 3-2-methyl-3-furyl thio heptan-4-one, 4-heptanone, 3-2-methyl-3-furanyl thio, 4-heptanone, 3-2-methyl-3-furyl thio, 3-2-methylfuran-3-yl sulfanylheptan-4-one, 3-2-methyl-3-furanyl thio-4-heptanone | |
3-(2-methylfuran-3-yl)sulfanylheptan-4-one |
Specifications
61295-41-8 | |
1.5015 | |
3-2-methyl-3-furylthio-4-heptanone, 3-2-methylfuran-3-yl thio heptan-4-one, fema no. 3570, 3-2-methyl-3-furyl thio-4-heptanone, 1,3-diethylacetonyl 3-methyl-3-furyl sulfide, 3-2-methyl-3-furyl thio heptan-4-one, 4-heptanone, 3-2-methyl-3-furanyl thio, 4-heptanone, 3-2-methyl-3-furyl thio, 3-2-methylfuran-3-yl sulfanylheptan-4-one, 3-2-methyl-3-furanyl thio-4-heptanone | |
CCCC(=O)C(CC)SC1=C(C)OC=C1 | |
226.33 | |
226.34 | |
3-(2-Methyl-3-furylthio)-4-heptanone |
C12H18O2S | |
MFCD01723735 | |
GFRRQSASJZMMJC-UHFFFAOYNA-N | |
3-(2-methylfuran-3-yl)sulfanylheptan-4-one | |
43598 | |
97% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 262-690-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures