missing translation for 'onlineSavingsMsg'
Learn More

[3-(2-Morpholinoethoxy)phenyl]methylamine, 95%, Thermo Scientific™

£26.00 - £69.00

Chemical Identifiers

CAS 857284-08-3
Molecular Formula C13H20N2O2
Molecular Weight (g/mol) 236.315
MDL Number MFCD07772847
InChI Key KRJWFDTWNMICLY-UHFFFAOYSA-N
Synonym 3-2-morpholinoethoxy phenyl methylamine, 3-2-morpholin-4-yl ethoxy phenyl methanamine, 3-2-morpholin-4-yl-ethoxy-benzylamine, 3-2-morpholin-4-yl ethoxy phenyl methylamine, benzenemethanamine, 3-2-4-morpholinyl ethoxy, 1-3-2-morpholin-4-yl ethoxy phenyl methanamine, 3-2-morpholin-4-ylethoxy phenyl methanamine, 3-2-morpholinoethoxy phenylmethylamine, 3-morpholinoethoxy-phenyl methyl amin, 3-2-morpholinoethoxy phenyl methanamine
PubChem CID 7164624
IUPAC Name [3-(2-morpholin-4-ylethoxy)phenyl]methanamine
SMILES C1COCCN1CCOC2=CC=CC(=C2)CN
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10612045
SDS View Documents
Thermo Scientific Maybridge
CC42013CB
250 mg Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10209662
SDS View Documents
Thermo Scientific Maybridge
CC42013DA
1 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
[3-(2-Morpholinoethoxy)phenyl]methylamine, 95%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.