Learn More
DWK Life Sciences Kimble Chase™ 3.3 Borosilicate Glass Test Tubes
Use round-bottom 3.3 Borosilicate Glass Test Tubes for laboratory needs when heat resistance is critical. These disposable test tubes are thick-walled and autoclavable.
Brand:  DWK Life Sciences A610010000911
 View more versions of this product
Product Code. 11863380
  • £33.05 / Pack of 100
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Disposable
  • Borosilicate glass
  • Excellent resistance to temperature changes
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Round bottom
1Each
10 mm
100 mm
1/Pk.
Test Tube