Learn More
3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals
£16.68 - £153.00
Chemical Identifiers
CAS | 95-76-1 |
---|---|
MDL Number | MFCD00007768 |
InChI Key | SDYWXFYBZPNOFX-UHFFFAOYSA-N |
Synonym | 3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline |
PubChem CID | 7257 |
ChEBI | CHEBI:16767 |
IUPAC Name | 3,4-dichloroaniline |
SMILES | C1=CC(=C(C=C1N)Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10293990
|
Thermo Scientific Acros
113110050 |
5 g | Glass bottle |
£16.68
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10204000
|
Thermo Scientific Acros
113112500 |
250 g | Glass bottle |
£43.82
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10409410
|
Thermo Scientific Acros
113110010 |
1 kg | Glass bottle |
£153.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-76-1 | |
SDYWXFYBZPNOFX-UHFFFAOYSA-N | |
7257 | |
3,4-dichloroaniline |
MFCD00007768 | |
3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline | |
CHEBI:16767 | |
C1=CC(=C(C=C1N)Cl)Cl |
Specifications
95-76-1 | |
100.0 | |
Beige to Brown | |
166°C | |
97.5% min. (GC) | |
Cl2C6H3NH2 | |
12, 626 | |
3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline | |
SDYWXFYBZPNOFX-UHFFFAOYSA-N | |
3,4-dichloroaniline | |
7257 | |
162.02 | |
Crystalline Mass or Chunks |
97.5 | |
69.0°C to 73.0°C | |
272.0°C | |
Authentic | |
C6H5Cl2N | |
MFCD00007768 | |
15, 3063 | |
Solubility in water: 0.06g/100mL in water. Other solubilities: soluble in most common organic solvents,very soluble in alcohol and ether,slightly soluble in benzene | |
C1=CC(=C(C=C1N)Cl)Cl | |
162.02 | |
CHEBI:16767 | |
98% | |
3, 4-Dichloroaniline, 98% |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
May cause an allergic skin reaction.
Toxic in contact with skin.
Causes serious eye damage.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Gently wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-448-4
missing translation for 'rtecsNumber' : BX2625000
missing translation for 'tsca' : TSCA
Recommended Storage : Product darkens on storage
- 3,4-Dichloroaniline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only