missing translation for 'onlineSavingsMsg'
Learn More
3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals Product Code.: 10204000

3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals

Product Code. 10204000
250 g, Glass bottle
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10204000

Brand: Thermo Scientific Acros 113112500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-76-1
MDL Number MFCD00007768
InChI Key SDYWXFYBZPNOFX-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroanilineShow MoreShow Less
PubChem CID 7257
ChEBI CHEBI:16767
IUPAC Name 3,4-dichloroaniline
SMILES C1=CC(=C(C=C1N)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 69.0°C to 73.0°C
Color Beige to Brown
Boiling Point 272.0°C
Flash Point 166°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H5Cl2N
Linear Formula Cl2C6H3NH2
Quantity 250 g
Beilstein 12, 626
Merck Index 15, 3063
Molecular Weight (g/mol) 162.013
Formula Weight 162.02
Percent Purity 98%
Physical Form Crystalline Mass or Chunks
Chemical Name or Material 3, 4-Dichloroaniline, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442473 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number113110010,113110050,113112500
Lot NumberA0458931 Certificate TypeCertificate of Analysis Date05/01/2024 Catalog Number113110010,113110050,113112500
Lot NumberA0375623 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number113110010,113110050,113112500
Lot NumberA0442473 Certificate TypeCertificate of Analysis Date02/06/2022 Catalog Number113110010,113110050,113112500
Lot NumberA0409249 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number113110010,113110050,113112500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3,4-Dichloroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.