missing translation for 'onlineSavingsMsg'
Learn More
1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals Product Code.: 11457747

1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals

Product Code. 11457747
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11457747

Brand: Thermo Scientific Alfa Aesar A16290.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, aluminum and its alloys, some plastics.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-46-7
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000604
InChI Key OCJBOOLMMGQPQU-UHFFFAOYSA-N
Synonym p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evolaShow MoreShow Less
PubChem CID 4685
ChEBI CHEBI:28618
IUPAC Name 1,4-dichlorobenzene
SMILES C1=CC(=CC=C1Cl)Cl

Specifications

Melting Point 52°C to 56°C
Density 1.248
Boiling Point 173°C to 174°C
Flash Point 65°C (149°F)
Odor Strong
Quantity 500 g
UN Number UN3077
Beilstein 1680023
Merck Index 14,3057
Solubility Information It is insoluble in water.
Formula Weight 147
Percent Purity ≥99%
Chemical Name or Material 1,4-Dichlorobenzene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251659 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA16290.0E,A16290.36,A16290.22
Lot Number10250469 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberA16290.0E,A16290.36,A16290.22
Lot Number10220580 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog NumberA16290.0E,A16290.36,A16290.22
Lot Number10249005 Certificate TypeCertificate of Analysis Date13/12/2023 Catalog NumberA16290.0E,A16290.36,A16290.22
Lot Number10238486 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA16290.0E,A16290.36,A16290.22
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Dichlorobenzene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : p-Dichlorobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.