missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diamino-1,2,4-triazole, 98%, Thermo Scientific Chemicals

£20.40 - £177.89

Chemical Identifiers

CAS 1455-77-2
Molecular Formula C2H5N5
Molecular Weight (g/mol) 99.09
MDL Number MFCD00005233
InChI Key PKWIYNIDEDLDCJ-UHFFFAOYSA-N
Synonym guanazole, 3,5-diamino-1,2,4-triazole, 4h-1,2,4-triazole-3,5-diamine, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, s-triazole, 3,5-diamino, 1,2,4-triazolidine, 3,5-diimino, unii-i01twm5267, ccris 3745, 3-amino-1h-1,2,4-triazol-5-ylamine
PubChem CID 15078
ChEBI CHEBI:75425
IUPAC Name 1H-1,2,4-triazole-3,5-diamine
SMILES C1(=NC(=NN1)N)N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10511252
View Documents
Thermo Scientific Acros
112440050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10489210
View Documents
Thermo Scientific Acros
112440250
25 g Glass bottle
£67.23
25g
Please to purchase this item. Need a web account? Register with us today!
10167653
View Documents
Thermo Scientific Acros
112441000
100 g Glass bottle
£177.89
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1455-77-2
99.09
PKWIYNIDEDLDCJ-UHFFFAOYSA-N
15078
1H-1,2,4-triazole-3,5-diamine
C2H5N5
MFCD00005233
guanazole, 3,5-diamino-1,2,4-triazole, 4h-1,2,4-triazole-3,5-diamine, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, s-triazole, 3,5-diamino, 1,2,4-triazolidine, 3,5-diimino, unii-i01twm5267, ccris 3745, 3-amino-1h-1,2,4-triazol-5-ylamine
CHEBI:75425
C1(=NC(=NN1)N)N

Specifications

1455-77-2
100.0
White to Beige
98%
MFCD00005233
guanazole, 3,5-diamino-1,2,4-triazole, 4h-1,2,4-triazole-3,5-diamine, 3,5-diamino-1h-1,2,4-triazole, 3,5-diamino-s-triazole, s-triazole, 3,5-diamino, 1,2,4-triazolidine, 3,5-diimino, unii-i01twm5267, ccris 3745, 3-amino-1h-1,2,4-triazol-5-ylamine
C1(=NC(=NN1)N)N
99.09
CHEBI:75425
98%
3,5-Diamino-1,2,4-triazole
97.5
204°C to 208°C
Authentic
C2H5N5
26, 193
PKWIYNIDEDLDCJ-UHFFFAOYSA-N
1H-1,2,4-triazole-3,5-diamine
15078
99.09
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-937-2

Product Identifier
  • 3,5-Diamino-1,2,4-triazole

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.