missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromo-1H-1,2,4-triazole, 97%, Thermo Scientific™
£8.10 - £158.00
Chemical Identifiers
CAS | 7411-23-6 |
---|---|
Molecular Formula | C2HBr2N3 |
Molecular Weight (g/mol) | 226.859 |
InChI Key | FRAKFBWDPXYIQO-UHFFFAOYSA-N |
Synonym | 3,5-dibromo-4h-1,2,4-triazole, 3,5-dibromo-1,2,4-triazole, 1h-1,2,4-triazole, 3,5-dibromo, s-triazole, 3,5-dibromo, 3,5-dibromo-s-triazole, so, deblqxbskfssp@, pubchem13285, acmc-1bktu, 1h-1,4-triazole, 3,5-dibromo |
PubChem CID | 81904 |
IUPAC Name | 3,5-dibromo-1H-1,2,4-triazole |
SMILES | C1(=NC(=NN1)Br)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10041674
|
Thermo Scientific Maybridge
MO07857CB |
250 mg | Amber Glass Bottle |
£8.10
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10729595
|
Thermo Scientific Maybridge
MO07857DA |
1 g | Amber Glass Bottle |
£15.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10051094
|
Thermo Scientific Maybridge
MO07857EB |
25 g | Amber Glass Bottle | N/A | |||||
Description
Chemical Identifiers
7411-23-6 | |
226.859 | |
3,5-dibromo-4h-1,2,4-triazole, 3,5-dibromo-1,2,4-triazole, 1h-1,2,4-triazole, 3,5-dibromo, s-triazole, 3,5-dibromo, 3,5-dibromo-s-triazole, so, deblqxbskfssp@, pubchem13285, acmc-1bktu, 1h-1,4-triazole, 3,5-dibromo | |
3,5-dibromo-1H-1,2,4-triazole |
C2HBr2N3 | |
FRAKFBWDPXYIQO-UHFFFAOYSA-N | |
81904 | |
C1(=NC(=NN1)Br)Br |
Specifications
7411-23-6 | |
C2HBr2N3 | |
FRAKFBWDPXYIQO-UHFFFAOYSA-N | |
3,5-dibromo-1H-1,2,4-triazole | |
81904 | |
97% |
97% | |
3,5-dibromo-4h-1,2,4-triazole, 3,5-dibromo-1,2,4-triazole, 1h-1,2,4-triazole, 3,5-dibromo, s-triazole, 3,5-dibromo, 3,5-dibromo-s-triazole, so, deblqxbskfssp@, pubchem13285, acmc-1bktu, 1h-1,4-triazole, 3,5-dibromo | |
C1(=NC(=NN1)Br)Br | |
226.859 | |
226.86 | |
3,5-Dibromo-1H-1,2,4-triazole |