Learn More
3,5-Dibromobenzoic acid, 98%, Thermo Scientific™
£17.78 - £220.00
Chemical Identifiers
CAS | 618-58-6 |
---|---|
Molecular Formula | C7H4Br2O2 |
Molecular Weight (g/mol) | 279.92 |
MDL Number | MFCD00051758 |
InChI Key | SFTFNJZWZHASAQ-UHFFFAOYSA-N |
Synonym | benzoic acid, 3,5-dibromo, 3,5-dibromobenzoicacid, benzoicacid, 3,5-dibromo, buttpark 99\57-19, pubchem3861, acmc-209mwz, maybridge1_003318, 3,5-dibromo-benzoic acid, 4-09-00-01028 beilstein handbook reference, ksc358a3h |
PubChem CID | 12063 |
IUPAC Name | 3,5-dibromobenzoic acid |
SMILES | OC(=O)C1=CC(Br)=CC(Br)=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3,5-Dibromobenzoic acid may be used for the syntheses (+)-menthyl 3,5-dibromobenzoate, di-tert-butyl 4-[2-(tert-butoxycarbonyl)ethyl]-4-(3,5-dibromobenzamido)heptanedioate2, (L)-methyl 2-(3,5-dibromobenzamido)-3-phenylpropanoate.
Solubility
Very faint turbidity in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
618-58-6 | |
279.92 | |
SFTFNJZWZHASAQ-UHFFFAOYSA-N | |
12063 | |
OC(=O)C1=CC(Br)=CC(Br)=C1 |
C7H4Br2O2 | |
MFCD00051758 | |
benzoic acid, 3,5-dibromo, 3,5-dibromobenzoicacid, benzoicacid, 3,5-dibromo, buttpark 99\57-19, pubchem3861, acmc-209mwz, maybridge1_003318, 3,5-dibromo-benzoic acid, 4-09-00-01028 beilstein handbook reference, ksc358a3h | |
3,5-dibromobenzoic acid |
Specifications
618-58-6 | |
C7H4Br2O2 | |
1940691 | |
Very faint turbidity in methanol. | |
OC(=O)C1=CC(Br)=CC(Br)=C1 | |
279.92 | |
279.93 | |
3,5-Dibromobenzoic acid |
216°C to 224°C | |
MFCD00051758 | |
benzoic acid, 3,5-dibromo, 3,5-dibromobenzoicacid, benzoicacid, 3,5-dibromo, buttpark 99\57-19, pubchem3861, acmc-209mwz, maybridge1_003318, 3,5-dibromo-benzoic acid, 4-09-00-01028 beilstein handbook reference, ksc358a3h | |
SFTFNJZWZHASAQ-UHFFFAOYSA-N | |
3,5-dibromobenzoic acid | |
12063 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'rtecsNumber' : DG6290010
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only