Learn More
3,5-Dichloro-4-pyridone-1-acetic acid, 98%, Thermo Scientific™
£33.80 - £135.00
Chemical Identifiers
CAS | 56187-37-2 |
---|---|
Molecular Formula | C7H5Cl2NO3 |
Molecular Weight (g/mol) | 222.021 |
MDL Number | MFCD02180547 |
InChI Key | MFMXEEQESUEMSB-UHFFFAOYSA-N |
Synonym | 3,5-dichloro-4-pyridone-1-acetic acid, 3,5-dichloro-4-pyridone-n-acetic acid, 2-3,5-dichloro-4-oxopyridin-1 4h-yl acetic acid, 1 4h-pyridineacetic acid, 3,5-dichloro-4-oxo, 3,5-dichloro-4-oxo-4h-pyridine-1-acetic acid, 3,5-dichloro-4-oxopyridin-1-yl acetic acid, 3,5-dichloro-4-pyridine-n-acetic acid, 2-3,5-dichloro-4-oxohydropyridyl acetic acid, pubchem11174, ksc272s7b |
PubChem CID | 92052 |
IUPAC Name | 2-(3,5-dichloro-4-oxopyridin-1-yl)acetic acid |
SMILES | C1=C(C(=O)C(=CN1CC(=O)O)Cl)Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3,5-Dichloro-4-pyridone-1-acetic acid is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
56187-37-2 | |
222.021 | |
MFMXEEQESUEMSB-UHFFFAOYSA-N | |
92052 | |
C1=C(C(=O)C(=CN1CC(=O)O)Cl)Cl |
C7H5Cl2NO3 | |
MFCD02180547 | |
3,5-dichloro-4-pyridone-1-acetic acid, 3,5-dichloro-4-pyridone-n-acetic acid, 2-3,5-dichloro-4-oxopyridin-1 4h-yl acetic acid, 1 4h-pyridineacetic acid, 3,5-dichloro-4-oxo, 3,5-dichloro-4-oxo-4h-pyridine-1-acetic acid, 3,5-dichloro-4-oxopyridin-1-yl acetic acid, 3,5-dichloro-4-pyridine-n-acetic acid, 2-3,5-dichloro-4-oxohydropyridyl acetic acid, pubchem11174, ksc272s7b | |
2-(3,5-dichloro-4-oxopyridin-1-yl)acetic acid |
Specifications
56187-37-2 | |
C7H5Cl2NO3 | |
3,5-dichloro-4-pyridone-1-acetic acid, 3,5-dichloro-4-pyridone-n-acetic acid, 2-3,5-dichloro-4-oxopyridin-1 4h-yl acetic acid, 1 4h-pyridineacetic acid, 3,5-dichloro-4-oxo, 3,5-dichloro-4-oxo-4h-pyridine-1-acetic acid, 3,5-dichloro-4-oxopyridin-1-yl acetic acid, 3,5-dichloro-4-pyridine-n-acetic acid, 2-3,5-dichloro-4-oxohydropyridyl acetic acid, pubchem11174, ksc272s7b | |
MFMXEEQESUEMSB-UHFFFAOYSA-N | |
2-(3,5-dichloro-4-oxopyridin-1-yl)acetic acid | |
92052 | |
98% |
239°C to 241°C | |
MFCD02180547 | |
Slightly soluble in water. | |
C1=C(C(=O)C(=CN1CC(=O)O)Cl)Cl | |
222.021 | |
222.03 | |
3,5-Dichloro-4-pyridone-1-acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 260-040-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures