missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzoyl chloride, 98%, Thermo Scientific™

£37.17 - £331.00

Chemical Identifiers

CAS 17213-57-9
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.62
MDL Number MFCD00000676
InChI Key FTHPLWDYWAKYCY-UHFFFAOYSA-N
PubChem CID 87003
IUPAC Name 3,5-dimethoxybenzoyl chloride
SMILES COC1=CC(=CC(OC)=C1)C(Cl)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11497786
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14744.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11407796
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14744.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11417796
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14744.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Dimethoxybenzoyl chloride was used to study the mechanism and kinetics of solvolysis of 3,4- and 3,5-dimethoxybenzoyl chlorides in various binary solvents. 3,5-Dimethoxybenzoyl chloride undergoes addition reaction with 4,4-dimethyl-2-pentyne in presence of AlCl3 via 1,2- methyl shift. It is used as a chemical and organic intermediate.

Solubility
It hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.
Specifications

Chemical Identifiers

17213-57-9
200.62
FTHPLWDYWAKYCY-UHFFFAOYSA-N
3,5-dimethoxybenzoyl chloride
C9H9ClO3
MFCD00000676
87003
COC1=CC(=CC(OC)=C1)C(Cl)=O

Specifications

17213-57-9
152°C to 154°C (11 mmHg)
C9H9ClO3
UN3261
Moisture sensitive
It hydrolyzes in water.
COC1=CC(=CC(OC)=C1)C(Cl)=O
200.62
200.62
3,5-Dimethoxybenzoyl chloride
42°C to 44°C
>110°C (230°F)
MFCD00000676
511839
benzoyl chloride, 3,5-dimethoxy, 3,5-dimethoxy benzoyl chloride, 3,5-dimethoxy-benzoyl chloride, acmc-1c560, benzoyl chloride,3,5-dimethoxy, 3,5-dimethoxybenzoic acid chloride, 3,5-dimethoxybenzoyl chloride, 3,5-dimethoxy-1-benzenecarbonyl chloride
FTHPLWDYWAKYCY-UHFFFAOYSA-N
3,5-dimethoxybenzoyl chloride
87003
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 241-256-5

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications