Learn More
3,5-Dinitrobenzoyl Chloride, 99%, Thermo Scientific Chemicals
£27.60 - £276.00
Chemical Identifiers
CAS | 99-33-2 |
---|---|
Molecular Formula | C7H3ClN2O5 |
Molecular Weight (g/mol) | 230.56 |
MDL Number | MFCD00007248 |
InChI Key | NNOHXABAQAGKRZ-UHFFFAOYSA-N |
Synonym | benzoyl chloride, 3,5-dinitro, 3,5-dinitrobenzoic acid chloride, dnbc, 3,5-dinitrobenzoylchloride, unii-5jfa2dvm4d, ccris 3137, 5jfa2dvm4d, 3,5-dinitrobenzoyl, acmc-209scd, dnbc; |
PubChem CID | 7432 |
IUPAC Name | 3,5-dinitrobenzoyl chloride |
SMILES | C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10157700
|
Thermo Scientific Acros
408710250 |
25 g | Glass bottle |
£27.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10677021
|
Thermo Scientific Acros
408711000 |
100 g | Glass bottle |
£66.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10195313
|
Thermo Scientific Acros
408715000 |
500 g | Glass bottle |
£276.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-33-2 | |
230.56 | |
NNOHXABAQAGKRZ-UHFFFAOYSA-N | |
7432 | |
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)Cl |
C7H3ClN2O5 | |
MFCD00007248 | |
benzoyl chloride, 3,5-dinitro, 3,5-dinitrobenzoic acid chloride, dnbc, 3,5-dinitrobenzoylchloride, unii-5jfa2dvm4d, ccris 3137, 5jfa2dvm4d, 3,5-dinitrobenzoyl, acmc-209scd, dnbc; | |
3,5-dinitrobenzoyl chloride |
Specifications
99-33-2 | |
100.0 | |
Yellow to Beige | |
Authentic | |
C7H3ClN2O5 | |
09, 414 | |
15, 3303 | |
Solubility in water: decomposes | |
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)Cl | |
230.56 | |
230.56 | |
Crystals or Crystalline Needles |
98.5 | |
67.0°C to 70.0°C | |
196°C (15.0 hPa) | |
99% | |
MFCD00007248 | |
01,320 | |
benzoyl chloride, 3,5-dinitro, 3,5-dinitrobenzoic acid chloride, dnbc, 3,5-dinitrobenzoylchloride, unii-5jfa2dvm4d, ccris 3137, 5jfa2dvm4d, 3,5-dinitrobenzoyl, acmc-209scd, dnbc; | |
NNOHXABAQAGKRZ-UHFFFAOYSA-N | |
3,5-dinitrobenzoyl chloride | |
7432 | |
99% | |
3, 5-Dinitrobenzoyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-750-6
RUO – Research Use Only