missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Difluorophthalic acid, 98%, Thermo Scientific Chemicals
£58.37 - £180.00
Chemical Identifiers
CAS | 651-97-8 |
---|---|
Molecular Formula | C8H4F2O4 |
Molecular Weight (g/mol) | 202.113 |
MDL Number | MFCD03840499 |
InChI Key | VFLMWMTWWZGXGA-UHFFFAOYSA-N |
Synonym | 3,6-difluorobenzene-1,2-dicarboxylic acid, 1,4-difluorophthalic acid, 3,6-difluorophthalicacid, pubchem1945, acmc-1ays1, 3,6-difluoro-phthalic acid, ksc352e6d, 3,5-difluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 3,6-difluoro |
PubChem CID | 643385 |
IUPAC Name | 3,6-difluorophthalic acid |
SMILES | C1=CC(=C(C(=C1F)C(=O)O)C(=O)O)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15453377
|
Thermo Scientific Alfa Aesar
H26096.MD |
250 mg | |||||||
11382059
|
Thermo Scientific Alfa Aesar
H26096.03 |
1 g |
£180.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
651-97-8 | |
202.113 | |
VFLMWMTWWZGXGA-UHFFFAOYSA-N | |
643385 | |
C1=CC(=C(C(=C1F)C(=O)O)C(=O)O)F |
C8H4F2O4 | |
MFCD03840499 | |
3,6-difluorobenzene-1,2-dicarboxylic acid, 1,4-difluorophthalic acid, 3,6-difluorophthalicacid, pubchem1945, acmc-1ays1, 3,6-difluoro-phthalic acid, ksc352e6d, 3,5-difluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 3,6-difluoro | |
3,6-difluorophthalic acid |
Specifications
651-97-8 | |
C8H4F2O4 | |
3,6-difluorobenzene-1,2-dicarboxylic acid, 1,4-difluorophthalic acid, 3,6-difluorophthalicacid, pubchem1945, acmc-1ays1, 3,6-difluoro-phthalic acid, ksc352e6d, 3,5-difluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 3,6-difluoro | |
C1=CC(=C(C(=C1F)C(=O)O)C(=O)O)F | |
202.113 | |
202.11 | |
3,6-Difluorophthalic acid |
∼184°C (decomposition) | |
MFCD03840499 | |
VFLMWMTWWZGXGA-UHFFFAOYSA-N | |
3,6-difluorophthalic acid | |
643385 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only