missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Difluorophthalic anhydride, 97%, Thermo Scientific Chemicals
£28.89 - £83.60
Chemical Identifiers
CAS | 652-40-4 |
---|---|
Molecular Formula | C8H2F2O3 |
Molecular Weight (g/mol) | 184.098 |
MDL Number | MFCD00134537 |
InChI Key | AVLRPSLTCCWJKC-UHFFFAOYSA-N |
Synonym | 3,6-difluorophthalic anhydride, 4,7-difluoroisobenzofuran-1,3-dione, 3,6-difluorophthalicanhydride, 1,3-isobenzofurandione, 4,7-difluoro, 4,7-difluoro-isobenzofuran-1,3-dione, pubchem1947, 3,6-difluorophthalic anhydride treated bsa, acmc-1avp1, ksc914s6p, 3,6 difluoro phthalic anhydride |
PubChem CID | 513918 |
IUPAC Name | 4,7-difluoro-2-benzofuran-1,3-dione |
SMILES | C1=CC(=C2C(=C1F)C(=O)OC2=O)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15443427
|
Thermo Scientific Alfa Aesar
H26215.MC |
100 mg |
£28.89
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15453427
|
Thermo Scientific Alfa Aesar
H26215.ME |
500 mg | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
652-40-4 | |
184.098 | |
AVLRPSLTCCWJKC-UHFFFAOYSA-N | |
513918 | |
C1=CC(=C2C(=C1F)C(=O)OC2=O)F |
C8H2F2O3 | |
MFCD00134537 | |
3,6-difluorophthalic anhydride, 4,7-difluoroisobenzofuran-1,3-dione, 3,6-difluorophthalicanhydride, 1,3-isobenzofurandione, 4,7-difluoro, 4,7-difluoro-isobenzofuran-1,3-dione, pubchem1947, 3,6-difluorophthalic anhydride treated bsa, acmc-1avp1, ksc914s6p, 3,6 difluoro phthalic anhydride | |
4,7-difluoro-2-benzofuran-1,3-dione |
Specifications
652-40-4 | |
C8H2F2O3 | |
Moisture sensitive | |
AVLRPSLTCCWJKC-UHFFFAOYSA-N | |
4,7-difluoro-2-benzofuran-1,3-dione | |
513918 | |
97% |
218°C to 221°C | |
MFCD00134537 | |
3,6-difluorophthalic anhydride, 4,7-difluoroisobenzofuran-1,3-dione, 3,6-difluorophthalicanhydride, 1,3-isobenzofurandione, 4,7-difluoro, 4,7-difluoro-isobenzofuran-1,3-dione, pubchem1947, 3,6-difluorophthalic anhydride treated bsa, acmc-1avp1, ksc914s6p, 3,6 difluoro phthalic anhydride | |
C1=CC(=C2C(=C1F)C(=O)OC2=O)F | |
184.098 | |
184.1 | |
3,6-Difluorophthalic anhydride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only