Learn More
3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals
£40.89 - £426.00
Chemical Identifiers
CAS | 287944-16-5 |
---|---|
Molecular Formula | C11H19BO3 |
Molecular Weight (g/mol) | 210.08 |
MDL Number | MFCD11052631 |
InChI Key | DOSGEBYQRMBTGS-UHFFFAOYSA-N |
Synonym | 3,6-dihydro-2h-pyran-4-boronic acid pinacol ester, 2-3,6-dihydro-2h-pyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-pyran, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 2h-pyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 3,6-dihydro-2h-pyran-4-boronic acid, pinacol ester, 3,6-dihydro-2h-pyran-4-boronicacidpinacolester, 3,6-dihydro-2h-pyran-4-boronic acid pinacolester, 3,6 dihydro-2h-pyran-4-boronic acid pinacol ester |
PubChem CID | 11218053 |
IUPAC Name | 2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | CC1(C)OB(OC1(C)C)C1=CCOCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488926
|
Thermo Scientific Alfa Aesar
H64452.MD |
250 mg | |||||||
15468926
|
Thermo Scientific Alfa Aesar
H64452.03 |
1 g |
£136.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15478926
|
Thermo Scientific Alfa Aesar
H64452.06 |
5 g |
£426.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It plays an important role as intermediate for organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt plays an important role as intermediate for organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
Chemical Identifiers
287944-16-5 | |
210.08 | |
DOSGEBYQRMBTGS-UHFFFAOYSA-N | |
11218053 | |
CC1(C)OB(OC1(C)C)C1=CCOCC1 |
C11H19BO3 | |
MFCD11052631 | |
3,6-dihydro-2h-pyran-4-boronic acid pinacol ester, 2-3,6-dihydro-2h-pyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-pyran, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 2h-pyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 3,6-dihydro-2h-pyran-4-boronic acid, pinacol ester, 3,6-dihydro-2h-pyran-4-boronicacidpinacolester, 3,6-dihydro-2h-pyran-4-boronic acid pinacolester, 3,6 dihydro-2h-pyran-4-boronic acid pinacol ester | |
2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
287944-16-5 | |
White | |
MFCD11052631 | |
Moisture sensitive | |
Slightly soluble in water. | |
CC1(C)OB(OC1(C)C)C1=CCOCC1 | |
210.08 | |
210.08 | |
Solid |
63°C to 67°C | |
C11H19BO3 | |
8548328 | |
3,6-dihydro-2h-pyran-4-boronic acid pinacol ester, 2-3,6-dihydro-2h-pyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-pyran, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 2h-pyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyran, 3,6-dihydro-2h-pyran-4-boronic acid, pinacol ester, 3,6-dihydro-2h-pyran-4-boronicacidpinacolester, 3,6-dihydro-2h-pyran-4-boronic acid pinacolester, 3,6 dihydro-2h-pyran-4-boronic acid pinacol ester | |
DOSGEBYQRMBTGS-UHFFFAOYSA-N | |
2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
11218053 | |
98% | |
3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only