missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Acetoxybenzaldehyde, 97%, Thermo Scientific™
£25.60 - £76.50
Chemical Identifiers
CAS | 34231-78-2 |
---|---|
Molecular Formula | C9H8O3 |
Molecular Weight (g/mol) | 164.16 |
MDL Number | MFCD00016603 |
InChI Key | GVUMZPWBUAGJBP-UHFFFAOYSA-N |
Synonym | 3-acetoxybenzaldehyde, benzaldehyde, 3-acetyloxy, acetoxybenzaldehyde, acetic acid 3-formylphenyl ester, m-acetoxybenzaldehyde, m-ace-toxybenzaldehyde, meta-acetoxybenzaldehyde, pubchem13227, 3-formylphenyl acetate |
PubChem CID | 118618 |
IUPAC Name | (3-formylphenyl) acetate |
SMILES | CC(=O)OC1=CC=CC(=C1)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11323057
|
Thermo Scientific Alfa Aesar
L13431.03 |
1 g |
£25.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333057
|
Thermo Scientific Alfa Aesar
L13431.06 |
5 g |
£76.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3-Acetoxybenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions. This material may darken on storage.
Chemical Identifiers
34231-78-2 | |
164.16 | |
GVUMZPWBUAGJBP-UHFFFAOYSA-N | |
118618 | |
CC(=O)OC1=CC=CC(=C1)C=O |
C9H8O3 | |
MFCD00016603 | |
3-acetoxybenzaldehyde, benzaldehyde, 3-acetyloxy, acetoxybenzaldehyde, acetic acid 3-formylphenyl ester, m-acetoxybenzaldehyde, m-ace-toxybenzaldehyde, meta-acetoxybenzaldehyde, pubchem13227, 3-formylphenyl acetate | |
(3-formylphenyl) acetate |
Specifications
34231-78-2 | |
138°C (20 mmHg) | |
1.533 | |
1941499 | |
3-acetoxybenzaldehyde, benzaldehyde, 3-acetyloxy, acetoxybenzaldehyde, acetic acid 3-formylphenyl ester, m-acetoxybenzaldehyde, m-ace-toxybenzaldehyde, meta-acetoxybenzaldehyde, pubchem13227, 3-formylphenyl acetate | |
GVUMZPWBUAGJBP-UHFFFAOYSA-N | |
(3-formylphenyl) acetate | |
118618 | |
97% |
1.17 | |
C9H8O3 | |
MFCD00016603 | |
Air Sensitive | |
Insoluble in water. | |
CC(=O)OC1=CC=CC(=C1)C=O | |
164.16 | |
164.16 | |
3-Acetoxybenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 251-890-4
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures