missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-1-phenylbutane, 98%, Thermo Scientific Chemicals

£109.00 - £460.00

Chemical Identifiers

CAS 22374-89-6
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00008090
InChI Key WECUIGDEWBNQJJ-UHFFFAOYSA-N
Synonym 1-methyl-3-phenylpropylamine, 3-amino-1-phenylbutane, 2-amino-4-phenylbutane, 1-phenyl-3-aminobutane, 4-phenyl-2-aminobutane, 4-phenyl-2-butylamine, 1-phenyl-3-amino-butan, benzenepropanamine, .alpha.-methyl, alpha-methylbenzenepropanamine, propylamine, 1-methyl-3-phenyl
PubChem CID 31160
IUPAC Name 4-phenylbutan-2-amine
SMILES CC(CCC1=CC=CC=C1)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11433088
View Documents
Thermo Scientific Alfa Aesar
A18804.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11443088
View Documents
Thermo Scientific Alfa Aesar
A18804.36
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a chemical intermediate. 1-Methyl-3-phenylpropylamine is used in the synthesis of N-substituted derivatives of (1-methyl-3- phenylpropyl)amine.

Solubility
Slightly soluble in water. Soluble in chloroform and ethyl acetate.

Notes
Air Sensitive. Store away from air, oxidizing agents and carbon dioxide. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Chemical Identifiers

22374-89-6
149.237
WECUIGDEWBNQJJ-UHFFFAOYSA-N
31160
CC(CCC1=CC=CC=C1)N
C10H15N
MFCD00008090
1-methyl-3-phenylpropylamine, 3-amino-1-phenylbutane, 2-amino-4-phenylbutane, 1-phenyl-3-aminobutane, 4-phenyl-2-aminobutane, 4-phenyl-2-butylamine, 1-phenyl-3-amino-butan, benzenepropanamine, .alpha.-methyl, alpha-methylbenzenepropanamine, propylamine, 1-methyl-3-phenyl
4-phenylbutan-2-amine

Specifications

22374-89-6
0.928
97°C (206°F)
C10H15N
MFCD00008090
2413110
1-methyl-3-phenylpropylamine, 3-amino-1-phenylbutane, 2-amino-4-phenylbutane, 1-phenyl-3-aminobutane, 4-phenyl-2-aminobutane, 4-phenyl-2-butylamine, 1-phenyl-3-amino-butan, benzenepropanamine, .alpha.-methyl, alpha-methylbenzenepropanamine, propylamine, 1-methyl-3-phenyl
WECUIGDEWBNQJJ-UHFFFAOYSA-N
4-phenylbutan-2-amine
31160
98%
-50°C
220°C to 222°C
Amine-like
1.514
UN2922
Air Sensitive
Slightly soluble in water. Soluble in chloroform and ethyl acetate.
CC(CCC1=CC=CC=C1)N
149.237
149.24
3-Amino-1-phenylbutane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

missing translation for 'einecsNumber' : 244-942-2

missing translation for 'rtecsNumber' : UI3920000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.