Learn More
3-Amino-4-iodo-1H-indazole, 95%, Thermo Scientific™
£69.70 - £117.00
Chemical Identifiers
CAS | 599191-73-8 |
---|---|
Molecular Formula | C7H6IN3 |
Molecular Weight (g/mol) | 259.05 |
MDL Number | MFCD03426697 |
InChI Key | OEQIVIYSUJXCFG-UHFFFAOYSA-N |
Synonym | 4-iodo-1h-indazol-3-ylamine, 3-amino-4-iodo-1h-indazole, 1h-indazol-3-amine, 4-iodo, 3-amino-4-iodoindazole, 4-iodo-1h-indazol-amine, ksc268o0n, 4-iodo-1h-indazole-3-amine, 4-iodo-1h-indazole-3-ylamine, 3-amino-4-iodo-indazole |
PubChem CID | 4057787 |
IUPAC Name | 4-iodo-1H-indazol-3-amine |
SMILES | NC1=NNC2=CC=CC(I)=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10311730
|
Thermo Scientific Acros
435750010 |
1 g | |||||||
10199359
|
Thermo Scientific Acros
435750050 |
5 g |
£117.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
599191-73-8 | |
259.05 | |
OEQIVIYSUJXCFG-UHFFFAOYSA-N | |
4057787 | |
NC1=NNC2=CC=CC(I)=C12 |
C7H6IN3 | |
MFCD03426697 | |
4-iodo-1h-indazol-3-ylamine, 3-amino-4-iodo-1h-indazole, 1h-indazol-3-amine, 4-iodo, 3-amino-4-iodoindazole, 4-iodo-1h-indazol-amine, ksc268o0n, 4-iodo-1h-indazole-3-amine, 4-iodo-1h-indazole-3-ylamine, 3-amino-4-iodo-indazole | |
4-iodo-1H-indazol-3-amine |
Specifications
599191-73-8 | |
100.0 | |
White to Brown | |
95% | |
C7H6IN3 | |
4-iodo-1h-indazol-3-ylamine, 3-amino-4-iodo-1h-indazole, 1h-indazol-3-amine, 4-iodo, 3-amino-4-iodoindazole, 4-iodo-1h-indazol-amine, ksc268o0n, 4-iodo-1h-indazole-3-amine, 4-iodo-1h-indazole-3-ylamine, 3-amino-4-iodo-indazole | |
NC1=NNC2=CC=CC(I)=C12 | |
259.05 | |
259.05 | |
Crystalline Solid |
94.0 | |
150°C to 155°C | |
Authentic | |
Glass bottle | |
MFCD03426697 | |
OEQIVIYSUJXCFG-UHFFFAOYSA-N | |
4-iodo-1H-indazol-3-amine | |
4057787 | |
≥94.0% | |
3-Amino-4-iodo-1H-indazole |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
- 3-Amino-4-iodo-1H-indazole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only