Learn More
3-Aminopropionitrile, 98%, stab. with potassium carbonate, Thermo Scientific Chemicals
£23.60 - £254.00
Chemical Identifiers
CAS | 151-18-8 |
---|---|
Molecular Formula | C3H6N2 |
Molecular Weight (g/mol) | 70.10 |
MDL Number | MFCD00014820 |
InChI Key | AGSPXMVUFBBBMO-UHFFFAOYSA-N |
Synonym | 3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino |
PubChem CID | 1647 |
ChEBI | CHEBI:27413 |
IUPAC Name | 3-aminopropanenitrile |
SMILES | NCCC#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11479973
|
Thermo Scientific Alfa Aesar
A13043.03 |
1 g |
£23.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489973
|
Thermo Scientific Alfa Aesar
A13043.06 |
5 g |
£65.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499973
|
Thermo Scientific Alfa Aesar
A13043.14 |
25 g |
£254.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.
Solubility
Soluble in water.
Notes
It is air and moisture sensitive. Incompatible with oxidizing agents, acids and heat.
Chemical Identifiers
151-18-8 | |
70.10 | |
AGSPXMVUFBBBMO-UHFFFAOYSA-N | |
1647 | |
3-aminopropanenitrile |
C3H6N2 | |
MFCD00014820 | |
3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino | |
CHEBI:27413 | |
NCCC#N |
Specifications
151-18-8 | |
79°C to 81°C (16 mmHg) | |
C3H6N2 | |
1.4375 | |
UN2735 | |
Air and moisture sensitive | |
3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino | |
AGSPXMVUFBBBMO-UHFFFAOYSA-N | |
3-aminopropanenitrile | |
1647 | |
70.1 | |
3-Aminopropionitrile, stabilized with potassium carbonate |
0.952 | |
Amine-like | |
H2NCH2CH2CN | |
MFCD00014820 | |
1698848 | |
14,469 | |
Soluble in water. | |
NCCC#N | |
70.10 | |
CHEBI:27413 | |
98% |
Safety and Handling
GHS H Statement
H301-H314-H318-H361-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
Combustible liquid.
P201-P202-P210-P235-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H314-H335-H361
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 205-786-0
missing translation for 'rtecsNumber' : UG0350000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only