missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(BOC-Aminomethyl)benzoic acid, 97%, Thermo Scientific™
£142.69 - £466.33
Chemical Identifiers
CAS | 117445-22-4 |
---|---|
Molecular Formula | C13H17NO4 |
Molecular Weight (g/mol) | 251.28 |
MDL Number | MFCD00273426 |
InChI Key | MQNHKLMHRZYTBZ-UHFFFAOYSA-N |
Synonym | boc-3-aminomethylbenzoic acid, 3-tert-butoxycarbonyl amino methyl benzoic acid, n-boc-3-aminomethyl-benzoic acid, 3-n-boc-aminomethyl benzoic acid, n-boc-3-aminomethylbenzoic acid, 3-boc-aminomethyl benzoic acid, boc-mamb, boc-3-amb-oh, 3-boc-aminomethyl-benzoic acid, n-boc-3-aminomethyl benzoic acid |
PubChem CID | 2755929 |
IUPAC Name | 3-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]benzoic acid |
SMILES | CC(C)(C)OC(=O)NCC1=CC=CC(=C1)C(O)=O |
Description
Chemical Identifiers
117445-22-4 | |
251.28 | |
MQNHKLMHRZYTBZ-UHFFFAOYSA-N | |
2755929 | |
CC(C)(C)OC(=O)NCC1=CC=CC(=C1)C(O)=O |
C13H17NO4 | |
MFCD00273426 | |
boc-3-aminomethylbenzoic acid, 3-tert-butoxycarbonyl amino methyl benzoic acid, n-boc-3-aminomethyl-benzoic acid, 3-n-boc-aminomethyl benzoic acid, n-boc-3-aminomethylbenzoic acid, 3-boc-aminomethyl benzoic acid, boc-mamb, boc-3-amb-oh, 3-boc-aminomethyl-benzoic acid, n-boc-3-aminomethyl benzoic acid | |
3-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]benzoic acid |
Specifications
129.0°C to 134.8°C | |
96.0 | |
White | |
C13H17NO4 | |
MFCD00273426 | |
boc-3-aminomethylbenzoic acid, 3-tert-butoxycarbonyl amino methyl benzoic acid, n-boc-3-aminomethyl-benzoic acid, 3-n-boc-aminomethyl benzoic acid, n-boc-3-aminomethylbenzoic acid, 3-boc-aminomethyl benzoic acid, boc-mamb, boc-3-amb-oh, 3-boc-aminomethyl-benzoic acid, n-boc-3-aminomethyl benzoic acid | |
CC(C)(C)OC(=O)NCC1=CC=CC(=C1)C(O)=O | |
251.28 | |
251.28 | |
Crystalline Powder or Flakes |
117445-22-4 | |
100.0 | |
96% min. (HPLC) | |
Authentic | |
Glass Bottle | |
MQNHKLMHRZYTBZ-UHFFFAOYSA-N | |
3-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]benzoic acid | |
2755929 | |
97% | |
3-(BOC-Aminomethyl)benzoic acid |
RUO – Research Use Only