missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-chlorochromone, 96%, Thermo Scientific™

£99.10

Chemical Identifiers

CAS 73220-38-9
Molecular Formula C9H4BrClO2
Molecular Weight (g/mol) 259.48
MDL Number MFCD02093123
InChI Key ZILSBPAUJJBEFF-UHFFFAOYSA-N
Synonym 3-bromo-6-chloro-4h-chromen-4-one, 3-bromo-6-chlorochromone, 4h-1-benzopyran-4-one,3-bromo-6-chloro, 3-bromo-6-chloro chromone, 3-bromo-6-chloro-4-oxo-4h-1-benzopyran, 3-bromo-6-chloro-4h-1-benzopyran-4-one
PubChem CID 3498945
IUPAC Name 3-bromo-6-chlorochromen-4-one
SMILES ClC1=CC=C2OC=C(Br)C(=O)C2=C1
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15474717
View Documents
Thermo Scientific Alfa Aesar
H32041.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

73220-38-9
259.48
ZILSBPAUJJBEFF-UHFFFAOYSA-N
3498945
ClC1=CC=C2OC=C(Br)C(=O)C2=C1
C9H4BrClO2
MFCD02093123
3-bromo-6-chloro-4h-chromen-4-one, 3-bromo-6-chlorochromone, 4h-1-benzopyran-4-one,3-bromo-6-chloro, 3-bromo-6-chloro chromone, 3-bromo-6-chloro-4-oxo-4h-1-benzopyran, 3-bromo-6-chloro-4h-1-benzopyran-4-one
3-bromo-6-chlorochromen-4-one

Specifications

73220-38-9
C9H4BrClO2
3-bromo-6-chloro-4h-chromen-4-one, 3-bromo-6-chlorochromone, 4h-1-benzopyran-4-one,3-bromo-6-chloro, 3-bromo-6-chloro chromone, 3-bromo-6-chloro-4-oxo-4h-1-benzopyran, 3-bromo-6-chloro-4h-1-benzopyran-4-one
ClC1=CC=C2OC=C(Br)C(=O)C2=C1
259.48
259.49
3-Bromo-6-chlorochromone
128°C to 130°C
MFCD02093123
ZILSBPAUJJBEFF-UHFFFAOYSA-N
3-bromo-6-chlorochromen-4-one
3498945
96%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications