missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofuran, 98%, Thermo Scientific Chemicals

£70.40 - £213.00

Chemical Identifiers

CAS 22037-28-1
Molecular Formula C4H3BrO
Molecular Weight (g/mol) 146.97
MDL Number MFCD00005347
InChI Key LXWLEQZDXOQZGW-UHFFFAOYSA-N
Synonym furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
PubChem CID 89164
IUPAC Name 3-bromofuran
SMILES BrC1=COC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10205470
View Documents Promotion Details
Thermo Scientific Acros
175260025
2.5 mL Glass bottle
£70.40
2.50mL
Please to purchase this item. Need a web account? Register with us today!
10474755
View Documents Promotion Details
Thermo Scientific Acros
175260100
10 mL Glass bottle
£213.00
10mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22037-28-1
146.97
LXWLEQZDXOQZGW-UHFFFAOYSA-N
89164
BrC1=COC=C1
C4H3BrO
MFCD00005347
furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
3-bromofuran

Specifications

22037-28-1
100.0
103.0°C (750.0 mmHg)
Authentic
C4H3BrO
MFCD00005347
furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
BrC1=COC=C1
copper
89164
98%
3-Bromofuran, Stabilized
97.5
1.6350g/mL
3°C
97.5% min. (GC)
1.4960 to 1.4980
1.635
LXWLEQZDXOQZGW-UHFFFAOYSA-N
3-bromofuran
146.97
146.97
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 244-744-6

Recommended Storage : −20°C

Product Identifier
  • 3-Bromofuran

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only