Learn More
3-Bromopropionic acid, 97%, Thermo Scientific Chemicals
£20.30 - £137.00
Chemical Identifiers
CAS | 590-92-1 |
---|---|
Molecular Formula | C3H5BrO2 |
Molecular Weight (g/mol) | 152.98 |
MDL Number | MFCD00002763 |
InChI Key | DHXNZYCXMFBMHE-UHFFFAOYSA-N |
Synonym | 3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r |
PubChem CID | 11553 |
IUPAC Name | 3-bromopropanoic acid |
SMILES | OC(=O)CCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11389509
|
Thermo Scientific Alfa Aesar
A11573.14 |
25 g |
£20.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484633
|
Thermo Scientific Alfa Aesar
A11573.22 |
100 g |
£30.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494633
|
Thermo Scientific Alfa Aesar
A11573.36 |
500 g |
£137.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Bromopropionic acid used as quaternization agent in amperometric biosensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Bromopropionic acid used as quaternization agent in amperometric biosensors.
Solubility
Soluble in water, alcohol, ether, chloroform, benzene
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
590-92-1 | |
152.98 | |
DHXNZYCXMFBMHE-UHFFFAOYSA-N | |
11553 | |
OC(=O)CCBr |
C3H5BrO2 | |
MFCD00002763 | |
3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r | |
3-bromopropanoic acid |
Specifications
590-92-1 | |
1.48 | |
65°C (149°F) | |
MFCD00002763 | |
1071333 | |
3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r | |
DHXNZYCXMFBMHE-UHFFFAOYSA-N | |
3-bromopropanoic acid | |
11553 | |
97% |
57°C to 63°C | |
140°C to 142°C (45 mmHg) | |
C3H5BrO2 | |
UN3261 | |
14,1433 | |
Soluble in water, alcohol, ether, chloroform, benzene | |
OC(=O)CCBr | |
152.98 | |
152.98 | |
3-Bromopropionic acid |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 209-694-1
missing translation for 'rtecsNumber' : UE7875000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only