missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyridine, 98+%, Thermo Scientific Chemicals

£36.78 - £472.35

Chemical Identifiers

CAS 626-55-1
Molecular Formula C5H4BrN
Molecular Weight (g/mol) 158.00
MDL Number MFCD00006373
InChI Key NYPYPOZNGOXYSU-UHFFFAOYSA-N
Synonym 3-pyridyl bromide, pyridine, 3-bromo, 3-bromo pyridine, 3-bromo-pyridine, 5-bromopyridine, unii-xmn8h2xe9h, 5-bromo pyridine, xmn8h2xe9h, 3-pyridylbromide, 3-bromopryidine
PubChem CID 12286
ChEBI CHEBI:51575
IUPAC Name 3-bromopyridine
SMILES BrC1=CC=CN=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11495333
SDS View Documents
Thermo Scientific Alfa Aesar
A12894.14
25 g
£36.78
25g
Please to purchase this item. Need a web account? Register with us today!
11405343
SDS View Documents
Thermo Scientific Alfa Aesar
A12894.22
100 g
£115.58
100g
Please to purchase this item. Need a web account? Register with us today!
11415343
SDS View Documents
Thermo Scientific Alfa Aesar
A12894.36
500 g
£472.35
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

3-Bromopyridine undergoes lithiation with LDA at low temperatures in the 4-position: Heterocycles, involved in formation of the Grignard by Mg exchange with i-PrMgCl and in transmetallation of the lithiated derivative with zinc chloride followed by treatment with an electrophile as a route to 4-substituted 3-bromopyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromopyridine undergoes lithiation with LDA at low temperatures in the 4-position: Heterocycles, involved in formation of the Grignard by Mg exchange with i-PrMgCl and in transmetallation of the lithiated derivative with zinc chloride followed by treatment with an electrophile as a route to 4-substituted 3-bromopyridines.

Solubility
Soluble in water (31 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong acids.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H227-H302-H332-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H301+H311+H331-H315-H319-H335

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 210-952-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromopyridine, 98+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.